missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyrylcholine Iodide, TCI America™

$25.97 - $116.11

Chemical Identifiers

CAS 2494-56-6
Molecular Formula C9H20INO2
Molecular Weight (g/mol) 301.168
MDL Number MFCD00038730
InChI Key GALNBQVDMJRFGJ-UHFFFAOYSA-M
PubChem CID 17232
IUPAC Name 2-butanoyloxyethyl(trimethyl)azanium;iodide
SMILES CCCC(=O)OCC[N+](C)(C)C.[I-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B07721G View Documents TCI America
B07721G
1g Each for $25.97
 
B077225G View Documents TCI America
B077225G
25g Each for $116.11
 
Specifications

Chemical Identifiers

2494-56-6
301.168
GALNBQVDMJRFGJ-UHFFFAOYSA-M
2-butanoyloxyethyl(trimethyl)azanium;iodide
C9H20INO2
MFCD00038730
17232
CCCC(=O)OCC[N+](C)(C)C.[I-]

Specifications

White-Yellow
1g
C9H20INO2
GALNBQVDMJRFGJ-UHFFFAOYSA-M
2-butanoyloxyethyl(trimethyl)azanium;iodide
17232
Crystal-Powder at 20°C
Butyrylcholine Iodide
86°C
2494-56-6
MFCD00038730
CCCC(=O)OCC[N+](C)(C)C.[I-]
301.168
301.17
≥99.0% (T)
Safety and Handling

Safety and Handling

RTECSNumber : BP0475000

TSCA : Yes