Learn More
Butyronitrile, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L02999AK
Description
Butyronitrile serves as an intermediate in the chemical and pharmaceutical industries. It is also involved in the preparation of n-butylamine, butanamide and butyric acid. It acts as a precursor to the poultry drug amprolium. Further, it reacts with phosphorochloridic acid diethyl ester to prepare (1-cyano-propyl)-phosphonic acid diethyl ester. It is also employed to study the effect of stereochemical factors on intervalence charge transfer. In addition to this, it is used in electrolyte composition in dye-sensitized solar cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsButyronitrile serves as an intermediate in the chemical and pharmaceutical industries. It is also involved in the preparation of n-butylamine, butanamide and butyric acid. It acts as a precursor to the poultry drug amprolium. Further, it reacts with phosphorochloridic acid diethyl ester to prepare (1-cyano-propyl)-phosphonic acid diethyl ester. It is also employed to study the effect of stereochemical factors on intervalence charge transfer. In addition to this, it is used in electrolyte composition in dye-sensitized solar cells.
Solubility
Miscible with benzene, alcohol, ether and dimethylformamide. Slightly soluble in water.
Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Specifications
Butyronitrile | |
-112°C | |
115°C to 117°C | |
C4H7N | |
CH3CH2CH2C≡N | |
250 mL | |
1361452 | |
butyronitrile, n-butyronitrile, propyl cyanide, 1-cyanopropane, n-propyl cyanide, butyrylonitrile, butyric acid nitrile, n-butanenitrile, propylkyanid, butane nitrile | |
KVNRLNFWIYMESJ-UHFFFAOYSA-N | |
butanenitrile | |
8008 | |
69.11 |
109-74-0 | |
0.792 | |
18°C (64°F) | |
1.384 | |
MFCD00001968 | |
UN2411 | |
14,1597 | |
Miscible with benzene,alcohol,ether and dimethylformamide. Slightly soluble in water. | |
CCCC#N | |
69.107 | |
CHEBI:51937 | |
99% |
Chemical Identifiers
109-74-0 | |
69.107 | |
KVNRLNFWIYMESJ-UHFFFAOYSA-N | |
8008 | |
butanenitrile |
C4H7N | |
MFCD00001968 | |
butyronitrile, n-butyronitrile, propyl cyanide, 1-cyanopropane, n-propyl cyanide, butyrylonitrile, butyric acid nitrile, n-butanenitrile, propylkyanid, butane nitrile | |
CHEBI:51937 | |
CCCC#N |
Safety and Handling
GHS H Statement
H225-H301-H311-H331
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P362-P372-P374-P380-P501c
H225-H300-H311+H331
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYRONITRILE
EINECSNumber : 203-700-6
RTECSNumber : ET8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only