Please login to your online account to display your discounted pricing

Alfa Aesar™ Butyric acid hydrazide, 95%

$97.93 - $97.93

Chemical Identifiers

Chemical Name or Material Butyric acid hydrazide
CAS 3538-65-6
Molecular Formula CH3CH2CH2CONHNH2
Formula Weight 102.14g/mol
MDL Number MFCD00025135
Beilstein 635863
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0197106

 
alfa aesar™
L01971-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0197114

 
alfa aesar™
L01971-14
25g Each for $97.93
Description & Specifications

Specifications

Quantity 5g
Boiling Point 120°C (10mmHg)
Melting Point 41° to 46°C
Quantity 5g
Chemical Name or Material Butyric acid hydrazide
CAS 3538-65-6
Assay 95%
Molecular Formula CH3CH2CH2CONHNH2
Formula Weight 102.14g/mol
MDL Number MFCD00025135
Beilstein 635863
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.