Please login to your online account to display your discounted pricing

Alfa Aesar™ Butyramidoxime

$85.43 - $298.34

Chemical Identifiers

Chemical Name or Material Butyramidoxime
CAS 27620-10-6
Molecular Formula C4H10N2O
Formula Weight 102.14
MDL Number MFCD05663152
Synonym N-Hydroxybutyramidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5088803

  SDS
alfa aesar™
H5088803
1g Each for $298.34

AAH50888MD

  SDS
alfa aesar™
H50888MD
250mg Each for $85.43
Description & Specifications

Specifications

Formula Weight 102.14
Chemical Name or Material Butyramidoxime
Molecular Formula C4H10N2O
MDL Number MFCD05663152
CAS 27620-10-6
Synonym N-Hydroxybutyramidine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.