missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Butyraldoxime, 98%

Manufacturer:  Alfa Aesar™ A1142022

 View more versions of this product

Catalog No. AAA1142022



Specifications

Specifications

Butyraldoxime
150°C to 152°C
100g
C4H9NO
MFCD00013943
1698799
KGGVGTQEGGOZRN-SNAWJCMRSA-N
(NE)-N-butylidenehydroxylamine
5324278
98%
58°C (136°F)
-29°C
110-69-0
CH3CH2CH2CH=NOH
UN2840
butyraldehyde oxime, butanal oxime, n-butyraldoxime, butylaldoxime, butanal, oxime, butyraldoxime, exkin 1, troykyd anti-skin bto, usaf am-6, m-butyraldehyde oxime
CCCC=NO
87.122
87.12

Chemical Identifiers

110-69-0
87.122
KGGVGTQEGGOZRN-SNAWJCMRSA-N
5324278
CCCC=NO
C4H9NO
MFCD00013943
butyraldehyde oxime, butanal oxime, n-butyraldoxime, butylaldoxime, butanal, oxime, butyraldoxime, exkin 1, troykyd anti-skin bto, usaf am-6, m-butyraldehyde oxime
(NE)-N-butylidenehydroxylamine
Safety and Handling

Safety and Handling

GHS H Statement
H311-H226-H302-H319
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYRALDOXIME

RTECSNumber : ES3500000

TSCA : TSCA

SDS Documents