Please login to your online account to display your discounted pricing

Alfa Aesar™ Butyraldoxime, 98%

Chemical Identifiers

Chemical Name or Material Butyraldoxime
CAS 110-69-0
Molecular Formula C4H9NO
Linear Formula CH3CH2CH2CH=NOH
Formula Weight 87.12
MDL Number MFCD00013943
UN Number UN2840
Beilstein 1698799
Synonym Butanal oxime; Butyraldehyde oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1142022

  SDS
alfa aesar™
A1142022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1142036

  SDS
alfa aesar™
A1142036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 87.12
Chemical Name or Material Butyraldoxime
Linear Formula CH3CH2CH2CH=NOH
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYRALDOXIME
Synonym Butanal oxime; Butyraldehyde oxime
Flash Point 58°C (136°F)
TSCA TSCA
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number ES3500000
UN Number UN2840
Assay 98%
MDL Number MFCD00013943
Health Hazard 1 Danger
Beilstein 1698799
Boiling Point 150° to 152°C
Molecular Formula C4H9NO
Melting Point −29°C
Quantity 100g
Health Hazard 2 GHS H Statement
H311-H226-H302-H319
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
CAS 110-69-0
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H226-H302-H319
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.