missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyraldehyde diethylacetal, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 393430250

 View more versions of this product

Catalog No. AC393430250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Butyraldehyde diethylacetal
0.8300g/mL
30°C
96% min. (GC)
C8H18O2
CH3CH2CH2CH(OCH2CH3)2
25g
butane, 1,1-diethoxy, butyraldehyde, diethyl acetal, butyraldehyde diethyl acetal, butylaldehyde diethyl acetal, n-butyraldehyde diethyl acetal, butyraldehyde diethyl acetal, analytical standard
UVHXZFGCCJLFMX-UHFFFAOYSA-N
1,1-diethoxybutane
77225
97%
3658-95-5
143.0°C
Authentic
Glass bottle
1.3950 to 1.3970 (20°C, 589nm)
MFCD00038316
0.83
Solubility in water: practically insoluble
CCCC(OCC)OCC
146.23
146.23

Chemical Identifiers

3658-95-5
146.23
UVHXZFGCCJLFMX-UHFFFAOYSA-N
77225
CCCC(OCC)OCC
C8H18O2
MFCD00038316
butane, 1,1-diethoxy, butyraldehyde, diethyl acetal, butyraldehyde diethyl acetal, butylaldehyde diethyl acetal, n-butyraldehyde diethyl acetal, butyraldehyde diethyl acetal, analytical standard
1,1-diethoxybutane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 222-913-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only