missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Butyltin trichloride, 97%, ACROS Organics™
Chemical Identifiers
CAS | 1118-46-3 |
---|---|
Molecular Formula | C4H9Cl3Sn |
Molecular Weight (g/mol) | 282.16 |
MDL Number | MFCD00000515 |
InChI Key | YMLFYGFCXGNERH-UHFFFAOYSA-K |
Synonym | butyl trichloro stannane, butyltin trichloride, butyltrichlorotin, monobutyltin trichloride, monotributyltin trichloride, n-butyltin trichloride, stannane, butyltrichloro, stannane, trichlorobutyl, trichlorobutylstannane, trichlorobutyltin |
PubChem CID | 14236 |
IUPAC Name | butyl(trichloro)stannane |
SMILES | CCCC[Sn](Cl)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC191201000
|
Thermo Fisher Scientific
191201000 |
100g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1118-46-3 | |
282.16 | |
YMLFYGFCXGNERH-UHFFFAOYSA-K | |
14236 | |
CCCC[Sn](Cl)(Cl)Cl |
C4H9Cl3Sn | |
MFCD00000515 | |
butyl trichloro stannane, butyltin trichloride, butyltrichlorotin, monobutyltin trichloride, monotributyltin trichloride, n-butyltin trichloride, stannane, butyltrichloro, stannane, trichlorobutyl, trichlorobutylstannane, trichlorobutyltin | |
butyl(trichloro)stannane |
Specifications
1.6900g/mL | |
Authentic | |
1.5215 to 1.5245 | |
93.0°C (10.0 mmHg) | |
100g | |
97% | |
CH3(CH2)3SnCl3 | |
04, IV, 4346 | |
butyl trichloro stannane, butyltin trichloride, butyltrichlorotin, monobutyltin trichloride, monotributyltin trichloride, n-butyltin trichloride, stannane, butyltrichloro, stannane, trichlorobutyl, trichlorobutylstannane, trichlorobutyltin | |
YMLFYGFCXGNERH-UHFFFAOYSA-K | |
butyl(trichloro)stannane | |
14236 | |
Liquid | |
Butyltin trichloride, 97% |
81°C | |
Glass bottle | |
1.69 | |
Colorless to Yellow or Straw-Yellow | |
1118-46-3 | |
C4H9Cl3Sn | |
MFCD00000515 | |
15, 1594 | |
Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents | |
CCCC[Sn](Cl)(Cl)Cl | |
282.16 | |
282.16 | |
97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Corrosive to the respiratory tract.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 214-263-6
RTECSNumber : WH6780000
TSCA : TSCA