missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Butylated Hydroxytoluene, 99.5%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210116290
Specifications
Butylated hydroxytoluene | |
500g | |
99.5% | |
2,6-di-t-butyl-4-methylphenol, 2,6-di-tert-butyl-4-methylphenol, 2,6-di-tert-butyl-p-cresol, bht, butylated hydroxytoluene, butylhydroxytoluene, dbpc, dibunol, ionol, stavox | |
CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C | |
220.356 | |
CHEBI:34247 | |
Crystalline |
1.048g/mL at 20°C | |
128-37-0 | |
C15H24O | |
NLZUEZXRPGMBCV-UHFFFAOYSA-N | |
2,6-ditert-butyl-4-methylphenol | |
31404 | |
220.36 |
Chemical Identifiers
128-37-0 | |
220.356 | |
2,6-di-t-butyl-4-methylphenol, 2,6-di-tert-butyl-4-methylphenol, 2,6-di-tert-butyl-p-cresol, bht, butylated hydroxytoluene, butylhydroxytoluene, dbpc, dibunol, ionol, stavox | |
CHEBI:34247 | |
CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C |
C15H24O | |
NLZUEZXRPGMBCV-UHFFFAOYSA-N | |
31404 | |
2,6-ditert-butyl-4-methylphenol |