missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 235235000

 View more versions of this product

Catalog No. AC235235000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Butylated hydroxyanisole
25013-16-5
White to Yellow
113°C
95% min. (GC)
C11H16O2
500 g
3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy
MRBKEAMVRSLQPH-UHFFFAOYSA-N
2-tert-butyl-4-methoxyphenol
8456
180.24
Crystalline Flakes or Crystals
96%
48°C to 55°C
268°C
Authentic
Plastic bottle
MFCD01779059
15,1549
Solubility in water: insoluble. Other solubilities: freely soluble in alcohol,propylene glycol,,chlorogorm,ether and acetone,slightly soluble in benzene
CC(C)(C)C1=C(C=CC(=C1)OC)O
180.24
CHEBI:76358
96%

Chemical Identifiers

25013-16-5
180.24
MRBKEAMVRSLQPH-UHFFFAOYSA-N
8456
2-tert-butyl-4-methoxyphenol
C11H16O2
MFCD01779059
3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy
CHEBI:76358
CC(C)(C)C1=C(C=CC(=C1)OC)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED:

GHS Signal Word: Warning

EINECSNumber : 246-563-8

RTECSNumber : SL1945000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only