Learn More
Butyl p-toluenesulfonate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 380590250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Butyl p-toluenesulfonate | |
Brown to Yellow | |
170°C to 172°C (10.0 mmHg) | |
98.5% min. (GC) | |
C11H16O3S | |
CH3C6H4SO3(CH2)3CH3 | |
25 g | |
butyl p-toluenesulfonate, butyl tosylate, n-butyl p-toluenesulfonate, butyl p-methylbenzenesulfonate, butyl-p-toluenesulfonate, p-toluenesulfonic acid, butyl ester, benzenesulfonic acid, 4-methyl-, butyl ester, butyl toluene-4-sulphonate, p-toluenesulfonic acid n-butyl ester, p-toluene sulfonic acid, butyl ester | |
QYJXDIUNDMRLAO-UHFFFAOYSA-N | |
butyl 4-methylbenzenesulfonate | |
13066 | |
99% |
778-28-9 | |
1.1200g/mL | |
Authentic | |
Glass bottle | |
1.5030 to 1.505 | |
MFCD00027203 | |
1.12 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol | |
CCCCOS(=O)(=O)C1=CC=C(C)C=C1 | |
228.31 | |
228.31 | |
Liquid |
Chemical Identifiers
778-28-9 | |
228.31 | |
QYJXDIUNDMRLAO-UHFFFAOYSA-N | |
13066 | |
CCCCOS(=O)(=O)C1=CC=C(C)C=C1 |
C11H16O3S | |
MFCD00027203 | |
butyl p-toluenesulfonate, butyl tosylate, n-butyl p-toluenesulfonate, butyl p-methylbenzenesulfonate, butyl-p-toluenesulfonate, p-toluenesulfonic acid, butyl ester, benzenesulfonic acid, 4-methyl-, butyl ester, butyl toluene-4-sulphonate, p-toluenesulfonic acid n-butyl ester, p-toluene sulfonic acid, butyl ester | |
butyl 4-methylbenzenesulfonate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 212-295-5
RUO – Research Use Only