missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl n-Octanoate, TCI America™

Manufacturer:  TCI America O002925ML

 View more versions of this product

Catalog No. O002925ML



Specifications

Specifications

Butyl n-Octanoate
Yellow
589-75-3
MFCD00048918
PSXNDMJWRZYVTM-UHFFFAOYSA-N
butyl octanoate
11517
200.32
≥99.0% (GC)
245°C
25mL
C12H24O2
butyl caprylate, butyl n-octanoate, n-butylcaprylate, octanoic acid, butyl ester, n-butyl octanoate, caprylic acid n-butyl ester, n-caprylic acid n-butyl ester, butyl-caprylate, n-butyl n-octanoate, octanoic acid butyl ester
CCCCCCCC(=O)OCCCC
200.322
CHEBI:87380
Clear Liquid at 20°C

Chemical Identifiers

589-75-3
200.322
PSXNDMJWRZYVTM-UHFFFAOYSA-N
11517
butyl octanoate
C12H24O2
MFCD00048918
butyl caprylate, butyl n-octanoate, n-butylcaprylate, octanoic acid, butyl ester, n-butyl octanoate, caprylic acid n-butyl ester, n-caprylic acid n-butyl ester, butyl-caprylate, n-butyl n-octanoate, octanoic acid butyl ester
CHEBI:87380
CCCCCCCC(=O)OCCCC
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0782

TSCA : Yes

Documents