missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl Myristate, TCI America™

$93.20 - $756.27

Chemical Identifiers

CAS 110-36-1
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.484
MDL Number MFCD00015077
InChI Key DHAZIUXMHRHVMP-UHFFFAOYSA-N
Synonym Myristic Acid Butyl Ester
PubChem CID 8047
IUPAC Name butyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M047725ML View Documents TCI America
M047725ML
25mL Each for $93.20
 
M0477500ML View Documents TCI America
M0477500ML
500mL Each for $756.27
 
Specifications

Chemical Identifiers

110-36-1
284.484
DHAZIUXMHRHVMP-UHFFFAOYSA-N
8047
CCCCCCCCCCCCCC(=O)OCCCC
C18H36O2
MFCD00015077
Myristic Acid Butyl Ester
butyl tetradecanoate

Specifications

195°C
25mL
C18H36O2
Myristic Acid Butyl Ester
CCCCCCCCCCCCCC(=O)OCCCC
284.484
284.48
≥97.0% (GC)
Yellow
110-36-1
MFCD00015077
DHAZIUXMHRHVMP-UHFFFAOYSA-N
butyl tetradecanoate
8047
Clear Liquid at 20°C
Butyl Myristate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

RTECSNumber : XB8550000

TSCA : Yes