missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl Methacrylate (stabilized with HQ), TCI America™

Manufacturer:  TCI America M008125ML

 View more versions of this product

Catalog No. M008125ML



Specifications

Specifications

Butyl Methacrylate (stabilized with HQ)
Colorless
25mL
C8H14O2
2227
SOGAXMICEFXMKE-UHFFFAOYSA-N
butyl 2-methylprop-2-enoate
7354
Clear Liquid at 20°C
164°C
-25°C
97-88-1
MFCD00009444
butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate
CCCCOC(=O)C(=C)C
142.198
142.20
≥99.0% (GC)

Chemical Identifiers

97-88-1
142.198
SOGAXMICEFXMKE-UHFFFAOYSA-N
7354
CCCCOC(=O)C(=C)C
C8H14O2
MFCD00009444
butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate
butyl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1039

RTECSNumber : OZ3675000

TSCA : Yes

Documents