Please login to your online account to display your discounted pricing

Butyl methacrylate, 99%, stabilized, ACROS Organics™

$57.32 - $57.32

Chemical Identifiers

Chemical Name or Material Butyl methacrylate
Name Note Stabilized
CAS 97-88-1
CAS 150-76-5
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H14O2
Linear Formula H2C=C(CH3)CO2(CH2)3CH3
Formula Weight 142.2
MDL Number MFCD00009444
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC216620051

  SDS
thermo fisher scientific
AC216620051
Plastic Jerrycan 5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC216620010

  SDS
thermo fisher scientific
216620010
Glass bottle 1L Each for $57.32
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.5%
Formula Weight 142.2
Quantity 5L
Color Clear
Packaging Plastic Jerrycan
Density 0.89g/mL
Beilstein 02, III, 1286
MDL Number MFCD00009444
CAS 97-88-1
Chemical Name or Material Butyl methacrylate
Molecular Formula C8H14O2
Health Hazard 1 GHS Signal Word: Warning
Refractive Index 1.4225 to 1.4245 (20°C, 589nm)
Stabilizer 10 to 110ppm MEHQ
Linear Formula H2C=C(CH3)CO2(CH2)3CH3
CAS 150-76-5
Infrared Spectrum Authentic
Name Note Stabilized
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.