Learn More
Butyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 216620010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Butyl methacrylate | |
97-88-1,150-76-5 | |
0.8900g/mL | |
41°C | |
98.5% min. (GC) | |
C8H14O2 | |
H2C=C(CH3)CO2(CH2)3CH3 | |
1 L | |
0.89 | |
Solubility in water: 3g/L in water (20°C). Other solubilities: very soluble in ether and ethanol,miscible in ethyl acetate | |
CCCCOC(=O)C(C)=C | |
142.20 | |
7354 | |
142.2 | |
Liquid |
Stabilized | |
-75.0°C | |
160.0°C to 163.0°C | |
Authentic | |
Glass bottle | |
1.4225 to 1.4245 | |
MFCD00009444 | |
02, III, 1286 | |
butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate | |
SOGAXMICEFXMKE-UHFFFAOYSA-N | |
butyl 2-methylprop-2-enoate | |
10 to 110ppm MEHQ | |
1.1 mPa.s (20°C) | |
99% |
Chemical Identifiers
97-88-1 | |
142.20 | |
SOGAXMICEFXMKE-UHFFFAOYSA-N | |
7354 | |
CCCCOC(=O)C(C)=C |
C8H14O2 | |
MFCD00009444 | |
butyl methacrylate, n-butyl methacrylate, butylmethacrylate, 2-propenoic acid, 2-methyl-, butyl ester, butyl 2-methacrylate, butylmethacrylaat, 2-methyl-butylacrylat, butil metacrilato, 2-methyl-butylacrylaat, 2-methyl-butylacrylate | |
butyl 2-methylprop-2-enoate |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-615-1
RUO – Research Use Only