missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 97-87-0 |
---|---|
Molecular Formula | C8H16O2 |
Molecular Weight (g/mol) | 144.214 |
MDL Number | MFCD00048773 |
InChI Key | JSLCOZYBKYHZNL-UHFFFAOYSA-N |
Synonym | Isobutyric Acid Butyl Ester |
PubChem CID | 7353 |
IUPAC Name | butyl 2-methylpropanoate |
SMILES | CCCCOC(=O)C(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
I031525ML |
TCI America
I031525ML |
25mL |
Each for $39.72
|
|
I0315500ML |
TCI America
I0315500ML |
500mL |
Each for $236.81
|
|
Chemical Identifiers
97-87-0 | |
144.214 | |
JSLCOZYBKYHZNL-UHFFFAOYSA-N | |
7353 | |
CCCCOC(=O)C(C)C |
C8H16O2 | |
MFCD00048773 | |
Isobutyric Acid Butyl Ester | |
butyl 2-methylpropanoate |
Specifications
150°C | |
25mL | |
C8H16O2 | |
3272 | |
JSLCOZYBKYHZNL-UHFFFAOYSA-N | |
butyl 2-methylpropanoate | |
7353 | |
Clear Liquid at 20°C | |
Butyl Isobutyrate |
Colorless | |
97-87-0 | |
MFCD00048773 | |
Isobutyric Acid Butyl Ester | |
CCCCOC(=O)C(C)C | |
144.214 | |
144.21 | |
≥98.0% (GC) |
Safety and Handling
TSCA : No