missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Butyl Glove Box Gloves, 8 in. Port

Butyl glove box gloves have the highest resistance to permeation by gases and vapors, and an exceptional resistance to a broad range of toxic chemicals.

$449.35 - $656.27

Specifications

Color Black
Latex Free Yes
Material Butyl
Size 9.75
Sterility Non-sterile
View More Specs

Products 3
Catalog Number Mfr. No. Hand Thickness (English) Thickness (Metric) Price Quantity  
Catalog Number Mfr. No. Hand Thickness (English) Thickness (Metric) Price Quantity  
19-018-047
View Documents Promotion Details
Honeywell™
8B1532A9Q
Ambidextrous 15 mil 0.38 mm
Pair for $449.35
 
11-395-42A
View Documents Promotion Details
Honeywell™
8B15329Q
Specific 15 mil 0.38 mm
Pair for $524.04
 
11-395-42B
View Documents Promotion Details
Honeywell™
8B303210H
Specific 30 mil 0.76 mm
Pair for $656.27
 
Description

Description

Butyl glove box gloves have the highest resistance to permeation by gases and vapors, and an exceptional resistance to a broad range of toxic chemicals.

  • Provides highest permeation resistance to vapors and gases
  • Resistance to a broad range of toxic chemicals, making it suitable choice for critical applications
  • Highly resistant to ketones (MEK, MIBK, acetone) and esters (tricresyl phosphate, amyl acetate, ethyl acetate)
  • Soft and flexible feel, even at low temperatures, which reduces hand fatigue and enhances comfort
  • Solvent based, multi-dip manufacturing process provides multiple layers of polymer (like glove within glove)
Specifications

Specifications

Black
Butyl
Non-sterile
Smooth
20.3 cm
81.3 cm
20.3 cm
Yes
9.75
Nuclear, defense, pharmaceutical, biotech, semi-conductor and manufacturing industries
8 in.
32 in.
8 in.
SDS
missing translation for 'documents'

missing translation for 'documents'