Please login to your online account to display your discounted pricing

Butyl chloroformate, 98%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  180171000

 View more versions of this product

Catalog No. AC180171000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 136.58
Quantity 100mL
Color Clear
Beilstein 03, II, 11
EINECS Number 209-750-5
TSCA TSCA
Chemical Name or Material Butyl chloroformate
Name Note 98%
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H9ClO2
Linear Formula ClCO2(CH2)3CH3
MDL Number MFCD00000650
CAS 592-34-7
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Refractive Index 1.4110 to 1.4130 (20°C, 589nm)
Health Hazard 1 Danger

Specifications

Chemical Name or Material Butyl chloroformate
Name Note 98%
CAS 592-34-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H9ClO2
Linear Formula ClCO2(CH2)3CH3
Formula Weight 136.58
MDL Number MFCD00000650
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents