missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl chloroformate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 180170250

 View more versions of this product

Catalog No. AC180170250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Coupling reagent
Specifications

Specifications

Butyl chloroformate
592-34-7
142.0°C
Authentic
Glass bottle
1.4110 to 1.4130
MFCD00000650
03, II, 11
butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate
NRDQFWXVTPZZAZ-UHFFFAOYSA-N
butyl carbonochloridate
61140
136.58
98%
1.0600g/mL
39°C
97.5% min. (GC)
C5H9ClO2
ClCO2(CH2)3CH3
25mL
1.06
Solubility in water: decomposes in water. Other solubilities: soluble in butanol,ether,benzol,soluble in most common organic solvents
CCCCOC(=O)Cl
136.58
0.95 mPa.s (20°C)
98%

Chemical Identifiers

592-34-7
136.58
NRDQFWXVTPZZAZ-UHFFFAOYSA-N
61140
CCCCOC(=O)Cl
C5H9ClO2
MFCD00000650
butyl chloroformate, n-butyl chloroformate, butylchloroformate, carbonochloridic acid, butyl ester, chloroformic acid butyl ester, butyl chlorocarbonate, n-butyl chlorocarbonate, formic acid, chloro-, butyl ester, butyl-chloroformate, butylchloro-formate
butyl carbonochloridate
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-750-5

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only