missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl benzoate, +98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 296402500

 View more versions of this product

Catalog No. AC296402500


 

Specifications Description & Specifications

Specifications

Butyl benzoate
136-60-7
Oily Liquid
98+%
C11H14O2
C6H5COO(CH2)3CH3
MFCD00009439
Clear yellow
98+%
Authentic
≥98.0%
Glass bottle
1.4960 to 1.4980 (20°C, 589nm)
178.23
15, 1554
250g

Chemical Identifiers

136-60-7
C11H14O2
178.23
Butyl benzoate, 98+%
98+%
C6H5COO(CH2)3CH3
MFCD00009439
≥98.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 205-252-7

RTECSNumber : DG4925000

TSCA : TSCA