Please login to your online account to display your discounted pricing

Butyl benzoate, +98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  296400050

 View more versions of this product

Catalog No. AC296400050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Oily Liquid
Formula Weight 178.23
Quantity 5g
Color Clear yellow
EINECS Number 205-252-7
TSCA TSCA
RTECS Number DG4925000
Merck Index 15, 1554
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula C6H5COO(CH2)3CH3
CAS 136-60-7
Chemical Name or Material Butyl benzoate
MDL Number MFCD00009439
Assay Percent Range 98+%
Molecular Formula C11H14O2
Name Note 98+%
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material Butyl benzoate
Name Note 98+%
CAS 136-60-7
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C11H14O2
Linear Formula C6H5COO(CH2)3CH3
Formula Weight 178.23
MDL Number MFCD00009439
Accessories

Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents