Learn More
Butyl acrylate, 99+%, stabilized, Thermo Scientific Chemicals
$116.45 - $484.40
Chemical Identifiers
CAS | 141-32-2 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00009446 |
InChI Key | CQEYYJKEWSMYFG-UHFFFAOYSA-N |
Synonym | butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove |
PubChem CID | 8846 |
ChEBI | CHEBI:3245 |
IUPAC Name | butyl prop-2-enoate |
SMILES | CCCCOC(=O)C=C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC215860010
|
Thermo Scientific Chemicals
215860010 |
1 L | Glass Bottle |
Each for $116.45
|
|
||||
AC215860100
|
Thermo Scientific Chemicals
215860100 |
10 L | Plastic drum |
Each for $484.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-32-2 | |
128.17 | |
CQEYYJKEWSMYFG-UHFFFAOYSA-N | |
8846 | |
butyl prop-2-enoate |
C7H12O2 | |
MFCD00009446 | |
butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove | |
CHEBI:3245 | |
CCCCOC(=O)C=C |
Specifications
141-32-2,150-76-5 | |
-64.0°C | |
145.0°C | |
Authentic | |
Glass Bottle | |
1.4170 to 1.4190 | |
MFCD00009446 | |
02, II, 388 | |
15, 1541 | |
Solubility in water: 1.4g/L (20°C) | |
CCCCOC(=O)C=C | |
128.17 | |
8846 | |
CHEBI:3245 | |
99+% | |
Butyl acrylate |
0.01% max. (as acrylic acid) | |
0.8900g/mL | |
39°C | |
99% min. (GC) | |
C7H12O2 | |
H2C=CHCO2(CH2)3CH3 | |
1 L | |
0.89 | |
butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove | |
CQEYYJKEWSMYFG-UHFFFAOYSA-N | |
butyl prop-2-enoate | |
10 to 20ppm MEHQ | |
0.869 mPa.s (20°C) | |
128.17 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful to aquatic li
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to f
GHS Signal Word: Warning
EINECSNumber : 205-480-7
RUO – Research Use Only