missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl Acetate 99.0+%, TCI America™

$23.55 - $32.97

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym 1-butyl acetate, acetate de butyle, acetic acid butyl ester, acetic acid n-butyl ester, acetic acid, butyl ester, butyl ethanoate, butylacetat, n-butyl acetate, n-butyl ethanoate, n-butylacetate
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A002425ML View Documents Tci America
A002425ML
25mL Each for $23.55
 
A0024500ML View Documents Tci America
A0024500ML
500mL Each for $32.97
 
Specifications

Chemical Identifiers

123-86-4
116.16
DKPFZGUDAPQIHT-UHFFFAOYSA-N
31272
butyl acetate
C6H12O2
MFCD00009445
1-butyl acetate, acetate de butyle, acetic acid butyl ester, acetic acid n-butyl ester, acetic acid, butyl ester, butyl ethanoate, butylacetat, n-butyl acetate, n-butyl ethanoate, n-butylacetate
CHEBI:31328
CCCCOC(=O)C

Specifications

126°C
-78°C
123-86-4
MFCD00009445
1-butyl acetate, acetate de butyle, acetic acid butyl ester, acetic acid n-butyl ester, acetic acid, butyl ester, butyl ethanoate, butylacetat, n-butyl acetate, n-butyl ethanoate, n-butylacetate
CCCCOC(=O)C
116.16
CHEBI:31328
Clear Liquid at 20°C
Butyl Acetate
Colorless
25mL
C6H12O2
1123
DKPFZGUDAPQIHT-UHFFFAOYSA-N
butyl acetate
31272
116.16
≥99.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0731

RTECSNumber : AF7350000

TSCA : Yes