missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl 4-hydroxybenzoate, +99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC403570025

 View more versions of this product

Catalog No. AC403570025

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥99.0%
Crystalline Powder
White to almost white
68° to 71°C
Authentic
94-26-8
C11H14O3
Butyl 4-hydroxybenzoate
Glass bottle
194.23
2.5kg
1.28g/mL
15, 1587
99+%
MFCD00016478
OHC6H4CO2(CH2)3CH3
99+%

Chemical Identifiers

94-26-8
C11H14O3
194.23
Butyl 4-hydroxybenzoate, 99+%
99+%
OHC6H4CO2(CH2)3CH3
MFCD00016478
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-318-7

RTECSNumber : DH1980000

TSCA : TSCA