Please login to your online account to display your discounted pricing

Butyl 4-hydroxybenzoate, +99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  403571000

 View more versions of this product

Catalog No. AC403571000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 194.23
Quantity 100g
Color White to almost white
Merck Index 15, 1587
EINECS Number 202-318-7
TSCA TSCA
Melting Point 68° to 71°C
RTECS Number DH1980000
Chemical Name or Material Butyl 4-hydroxybenzoate
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C11H14O3
MDL Number MFCD00016478
CAS 94-26-8
Packaging Glass bottle
Linear Formula OHC6H4CO2(CH2)3CH3
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Butyl 4-hydroxybenzoate
Name Note 99+%
CAS 94-26-8
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C11H14O3
Linear Formula OHC6H4CO2(CH2)3CH3
Formula Weight 194.23
MDL Number MFCD00016478
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents