missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl 2-Methylvalerate, TCI America™

$48.89 - $331.53

Chemical Identifiers

CAS 6297-41-2
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
InChI Key FMVUGLMJUHFPSJ-UHFFFAOYSA-N
Synonym 2-Methylvaleric Acid Butyl Ester, Butyl 2-Methylpentanoate, 2-Methylpentanoic Acid Butyl Ester
PubChem CID 95539
IUPAC Name butyl 2-methylpentanoate
SMILES CCCCOC(=O)C(C)CCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B335925ML View Documents TCI America
B335925ML
25mL Each for $48.89
 
B3359500ML View Documents TCI America
B3359500ML
500mL Each for $331.53
 
Specifications

Chemical Identifiers

6297-41-2
172.268
2-Methylvaleric Acid Butyl Ester, Butyl 2-Methylpentanoate, 2-Methylpentanoic Acid Butyl Ester
butyl 2-methylpentanoate
C10H20O2
FMVUGLMJUHFPSJ-UHFFFAOYSA-N
95539
CCCCOC(=O)C(C)CCC

Specifications

Colorless
6297-41-2
2-Methylvaleric Acid Butyl Ester, Butyl 2-Methylpentanoate, 2-Methylpentanoic Acid Butyl Ester
CCCCOC(=O)C(C)CCC
172.268
172.27
≥98.0% (GC)
25mL
C10H20O2
FMVUGLMJUHFPSJ-UHFFFAOYSA-N
butyl 2-methylpentanoate
95539
Clear Liquid at 20°C
Butyl 2-Methylvalerate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0782

TSCA : Yes