Please login to your online account to display your discounted pricing

Butvar™ B-98, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC428482500

 View more versions of this product

Catalog No. AC428482500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Free-Flowing Powder
Quantity 250g
Color White
Assay 18.0 to 20.0% (Hydroxyl content) as PV alcohol, 2.5% max. (Acetate content) as PV acetate
Viscosity 6 to 9cPs (6% in methanol, 20°C)
Density 1.1g/mL
Volatiles 5.0 max.
Infrared Spectrum Authentic
Synonym Poly(vinyl butyral) resin
MDL Number MFCD00084458
Packaging Glass bottle
CAS 63148-65-2
Chemical Name or Material Butvar™ B-98
Additional Information Molecular weigth (by SEC): 40, 000 to 70, 000

Specifications

Chemical Name or Material Butvar™ B-98
Assay 18.0 to 20.0% (Hydroxyl content) as PV alcohol, 2.5% max. (Acetate content) as PV acetate
MDL Number MFCD00084458
Synonym Poly(vinyl butyral) resin