Please login to your online account to display your discounted pricing

Butoxycarbonylmethyl butyl phthalate, 95%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC375045000

 View more versions of this product

Catalog No. AC375045000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 336.38
Physical Form Liquid
Quantity 500mL
Color Clear colorless
Density 1.1000g/mL
Packaging Glass bottle
Refractive Index 1.4900 to 1.4920 (20°C, 589nm)
CAS 84-74-2
Health Hazard 1 Danger
Molecular Formula C18H24O6
CAS 85-70-1
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Butoxycarbonylmethyl butyl phthalate
Assay Percent Range 95%
Infrared Spectrum Authentic
MDL Number MFCD00053793

Specifications

Chemical Name or Material Butoxycarbonylmethyl butyl phthalate
CAS 85-70-1
CAS 84-74-2
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C18H24O6
Formula Weight 336.38
MDL Number MFCD00053793
Accessories

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.

SDS & Safety Documents