missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butoxycarbonylmethyl butyl phthalate, 95%, ACROS Organics™

Chemical Identifiers

CAS 85-70-1,84-74-2
Assay Percent Range 95%
Molecular Formula C18H24O6
Formula Weight 336.38
MDL Number MFCD00053793
Chemical Name or Material Butoxycarbonylmethyl butyl phthalate
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC375045000 SDS Thermo Fisher Scientific
AC375045000
Glass bottle 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

85-70-1,84-74-2
C18H24O6
MFCD00053793
≥94%
95%
336.38
Butoxycarbonylmethyl butyl phthalate

Specifications

85-70-1,84-74-2
Authentic
95%
C18H24O6
336.38
Clear colorless
Butoxycarbonylmethyl butyl phthalate
1.1000g/mL
Liquid
Glass bottle
1.4900 to 1.4920 (20°C, 589nm)
MFCD00053793
500mL
≥94%

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
May damage the unborn child. Suspected of damaging fertility.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger