Butane Micro Burner
A safe alternative to alcohol burners.
Manufacturer:  Fisher Scientific MF2001
Catalog No. S65148
Description
Description
A safe alternative to alcohol burners.
This small butane burner uses clean burning butane and features a trigger ignition, eliminating the need for wicks, matches or a central gas line.
  • Stable, tip-resistant design and easy-grip handle make the burner safe and portable
  • Flame height is fully adjustable
  • Maximum temperature of approximately 1300°C (2372°F)
  • Two-hour fuel capacity
  • Compatible with standard butane cartridges
Optional Accessories (Not Included):
  • Locking Tripod Stand (S65024)
  • Butane Refill (S64968)
Specifications
Specifications
Provides Safe, Portable Flame Applications in Any Work Area
3.5 in.
2372°F
Butane
Butane Micro Burner
8.89 cm
1300°C
Micro Burner