missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butadiene sulfone, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC107615000

 View more versions of this product

Catalog No. AC107615000


 

Specifications Description & Specifications

Specifications

Powder
63° to 67°C
500g
1.3000g/mL
17, V,1, 177
C4H6O2S
Butadiene sulfone
Plastic bottle
Authentic
≥97.5% (GC)
118.16
Yellow to orange or light brown
02,389; 03,272; 04,468; 12,455
2, 5-Dihydrothiophene-1, 1-dioxide
77-79-2
98%
MFCD00005481

Chemical Identifiers

77-79-2
C4H6O2S
MFCD00005481
Butadiene sulfone
98%
118.16
2, 5-Dihydrothiophene-1, 1-dioxide
≥97.5% (GC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.