missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Butadiene diepoxide, 95%

$46.61 - $120.69

Chemical Identifiers

CAS 1464-53-5
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00005120
InChI Key ZFIVKAOQEXOYFY-UHFFFAOYSA-N
Synonym 1,2:3,4-diepoxybutane, 1,3-butadiene diepoxide, 2,2'-bioxirane, bioxirane, butadiendioxyd, butadiene diepoxide, butadiene dioxide, butane diepoxide, diepoxybutane, dioxybutadiene
PubChem CID 11254
ChEBI CHEBI:23704
IUPAC Name 2-(oxiran-2-yl)oxirane
SMILES C1C(O1)C2CO2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0391903
View Documents
Alfa Aesar™
L0391903
1g Each for $46.61
 
AAL0391906
View Documents
Alfa Aesar™
L0391906
5g Each for $120.69
 
Description

Description

Applications
Butadiene diepoxide is used as a research chemical, as a chemical intermediate, as a curing agent for polymers, as a cross-linking agent for textile fabrics, and to prevent microbial spoilage. It is also been used to synthesize erythritol and pharmaceuticals.

Solubility
Soluble in water (1000 g/L).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1464-53-5
86.09
ZFIVKAOQEXOYFY-UHFFFAOYSA-N
11254
2-(oxiran-2-yl)oxirane
C4H6O2
MFCD00005120
1,2:3,4-diepoxybutane, 1,3-butadiene diepoxide, 2,2'-bioxirane, bioxirane, butadiendioxyd, butadiene diepoxide, butadiene dioxide, butane diepoxide, diepoxybutane, dioxybutadiene
CHEBI:23704
C1C(O1)C2CO2

Specifications

1.112
1.434
-19°C
1464-53-5
MFCD00005120
79831
1,2:3,4-diepoxybutane, 1,3-butadiene diepoxide, 2,2'-bioxirane, bioxirane, butadiendioxyd, butadiene diepoxide, butadiene dioxide, butane diepoxide, diepoxybutane, dioxybutadiene
ZFIVKAOQEXOYFY-UHFFFAOYSA-N
2-(oxiran-2-yl)oxirane
11254
86.09
Butadiene diepoxide
45°C (113°F)
56°C to 58°C (25mmHg)
1g
C4H6O2
UN2929
14,3676
Soluble in water (1000g/L).
C1C(O1)C2CO2
86.09
CHEBI:23704
95%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H330-H340-H350-H314-H226
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H301-H310+H330-H314-H335-H340-H350-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: I

EINECSNumber : 215-979-1

RTECSNumber : EJ8400000

TSCA : Yes

Recommended Storage : Ambient temperatures