Please login to your online account to display your discounted pricing

Butabindide oxalate, Tocris


CCK-inactivating serine protease inhibitor

Quantity 50mg

Manufacturer: r & d systems  132350

 View more versions of this product

Catalog No. 132350

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 393.44
Quantity 50mg
Chemical Name or Material Butabindide oxalate
Assay Percent Range >99%
Molecular Formula C17H25N3O2·C2H2O4
CAS 185213-03-0
Synonym (2S)-[1-[(2S)-2-amino-1-oxobutyl]-N-butyl]-2, 3-dihydro-1H-indole-2-carboxamide oxalate
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Butabindide oxalate
Assay Percent Range >99%
Molecular Formula C17H25N3O2·C2H2O4
Formula Weight 393.44
Synonym (2S)-[1-[(2S)-2-amino-1-oxobutyl]-N-butyl]-2, 3-dihydro-1H-indole-2-carboxamide oxalate
Accessories

Desiccate at -20°C