Please login to your online account to display your discounted pricing

Butabindide oxalate, Tocris

CCK-inactivating serine protease inhibitor

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material Butabindide oxalate
Assay Percent Range >99%
Molecular Formula C17H25N3O2·C2H2O4
Formula Weight 393.44
Synonym (2S)-[1-[(2S)-2-amino-1-oxobutyl]-N-butyl]-2, 3-dihydro-1H-indole-2-carboxamide oxalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

132350

 
r & d systems
132350
50mg Each for $976.15

132310

 
r & d systems
132310
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 393.44
Quantity 50mg
Chemical Name or Material Butabindide oxalate
Synonym (2S)-[1-[(2S)-2-amino-1-oxobutyl]-N-butyl]-2, 3-dihydro-1H-indole-2-carboxamide oxalate
Recommended Storage Desiccate at -20°C
Assay Percent Range >99%
Molecular Formula C17H25N3O2·C2H2O4
CAS 185213-03-0
Safety and Handling

Desiccate at -20°C