missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Busulfan, 98%

A cell cycle non-specific alkylating antineoplastic agent

$30.38 - $30.38

Chemical Identifiers

CAS 55-98-1
Molecular Formula C6H14O6S2
Molecular Weight (g/mol) 246.292
MDL Number MFCD00007562
InChI Key COVZYZSDYWQREU-UHFFFAOYSA-N
Synonym busulfan, myleran, busulphan, sulphabutin, leucosulfan, myelosan, busulfex, citosulfan, mielucin, misulban
PubChem CID 2478
ChEBI CHEBI:28901
IUPAC Name 4-methylsulfonyloxybutyl methanesulfonate
SMILES CS(=O)(=O)OCCCCOS(=O)(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6134806 View Documents Alfa Aesar™
J6134806
5g Each for $30.38
 
Specifications

Chemical Identifiers

55-98-1
246.292
COVZYZSDYWQREU-UHFFFAOYSA-N
2478
4-methylsulfonyloxybutyl methanesulfonate
C6H14O6S2
MFCD00007562
busulfan, myleran, busulphan, sulphabutin, leucosulfan, myelosan, busulfex, citosulfan, mielucin, misulban
CHEBI:28901
CS(=O)(=O)OCCCCOS(=O)(=O)C

Specifications

Brown
55-98-1
CH3SO2O(CH2)4OSO2CH3
UN2811
14,1505
COVZYZSDYWQREU-UHFFFAOYSA-N
4-methylsulfonyloxybutyl methanesulfonate
2478
246.29
98%
5g
C6H14O6S2
MFCD00007562
1791786
busulfan, myleran, busulphan, sulphabutin, leucosulfan, myelosan, busulfex, citosulfan, mielucin, misulban
CS(=O)(=O)OCCCCOS(=O)(=O)C
246.292
CHEBI:28901
Crystalline Powder
Busulfan
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H350
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause cancer.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : EK1750000