missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Burner Bunsen Meker: High Temperature
Ideal for lab work requiring a hotter flame than that attainable with a bunsen burner.
Manufacturer: Eisco™ CH0098FRSD
Description
- Features adjustable orifice for natural or cylinder gas
- Adjust the air supply with the shutter sleeve covering the air intake ports, needle valve controls gas flow
- An expanded Meker type top with Monel stabilizing and heat intensifying grid provides a short hot flame
- For 8 mm diameter tubing
- Grid top diameter approximately 42mm
Specifications
8 mm | |
4.3 in. | |
Natural Gas |
Lab work requiring a hotter flame than that attainable with a bunsen burner | |
215 mm | |
Meker Bunsen Burner |