GSC Go Science Crazy Burette and Pipette Brush
Brush and clean small containers like burettes or pipettes. GSC International Burette and Pipette Brushes employ soft, natural bristles to avoid scratching and galvanized handles to prevent rust.
Supplier: GSC Go Science Crazy 410360
Alert:
Intended for ages 13 and up.
Description
- Great for cleaning out burettes or pipettes in laboratory or classroom settings
- Small, natural bristles are soft and designed to clean small spaces without scratching
- Galvanized handles prevent rusting over repeated use
- Available in two sizes
Specifications
Soft | |
19 mm | |
760 mm |
Galvanized Steel | |
Cleaning burettes and pipettes | |
92 mm |