missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bupivacaine hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals

Inhibits TREK-1 channels and depolarizes the cell membrane

Supplier:  Thermo Scientific Chemicals J6283503

 View more versions of this product

Catalog No. AAJ6283503



Description

Description

It is employed as Na+ channel blocker, local anesthetic and cAMP production inhibitor. It acts as a surfactant molecule possessing both hydrophilic and lipophilic properties, adrenergic antagonist, EC 3.1.1.8 (cholinesterase) inhibitor and EC 3.6.3.8 (Ca(2+)-transporting ATPase) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as Na+ channel blocker, local anesthetic and cAMP production inhibitor. It acts as a surfactant molecule possessing both hydrophilic and lipophilic properties, adrenergic antagonist, EC 3.1.1.8 (cholinesterase) inhibitor and EC 3.6.3.8 (Ca(2+)-transporting ATPase) inhibitor.

Solubility
Soluble in water to 50 mM, in DMSO to 100 mM and in ethanol to 100 mM.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Bupivacaine hydrochloride monohydrate
255°C to 259°C
C18H31ClN2O2
1 g
14,1495
Soluble in water to 50 mM,in DMSO to 100 mM and in ethanol to 100 mM.
CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.O.Cl
342.908
342.9
Powder
73360-54-0
White
MFCD00078956
UN2811
bupivacaine hydrochloride monohydrate, lac-43, marcaine tn, +/--1-butyl-2',6'-pipecoloxylidide monohydrochloride, monohydrate, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrate hydrochloride, bupivacaine hydrate hydrochloride, bupivacaine hcl h2-o, 2-piperidinecarboxamide, 1-butyl-n-2,6-dimethylphenyl-, monohydrochloride, monohydrate, bupivacaine hydrochloride ep, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride hydrate
HUCIWBPMHXGLFM-UHFFFAOYSA-N
1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride
5282419
≥98%

Chemical Identifiers

73360-54-0
342.908
HUCIWBPMHXGLFM-UHFFFAOYSA-N
5282419
CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.O.Cl
C18H31ClN2O2
MFCD00078956
bupivacaine hydrochloride monohydrate, lac-43, marcaine tn, +/--1-butyl-2',6'-pipecoloxylidide monohydrochloride, monohydrate, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrate hydrochloride, bupivacaine hydrate hydrochloride, bupivacaine hcl h2-o, 2-piperidinecarboxamide, 1-butyl-n-2,6-dimethylphenyl-, monohydrochloride, monohydrate, bupivacaine hydrochloride ep, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride hydrate
1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c

H300+H310+H330

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 241-917-8

RTECSNumber : TK6125000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only