missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bumetanide, 98+%

Inhibitor of the sodium-potassium-chloride cotransporter

$150.68 - $586.00

Chemical Identifiers

CAS 28395-03-1
Molecular Formula C17H20N2O5S
Molecular Weight (g/mol) 364.416
MDL Number MFCD00078949
InChI Key MAEIEVLCKWDQJH-UHFFFAOYSA-N
Synonym bumetanide, 3-butylamino-4-phenoxy-5-sulfamoylbenzoic acid, bumex, burinex, fordiuran, lunetoron, fontego, segurex, bumetanida, bumetanidum
PubChem CID 2471
ChEBI CHEBI:3213
IUPAC Name 3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid
SMILES CCCCNC1=C(C(=CC(=C1)C(=O)O)S(=O)(=O)N)OC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6230203 View Documents Alfa Aesar™
J6230203
1g Each for $150.68
 
AAJ6230206 View Documents Alfa Aesar™
J6230206
5g Each for $586.00
 
Description

Description

Applications
Bumetanide has been observed to block the depolarization of GABA in cortical development studies. It is been used as a specific inhibitor of Na+-K+-2Cl- cotransporter NKCC1

Solubility
Soluble in ethanol (10 mg/ml), DMSO (25 mg/ml), acetone, benzene, methanol, propylene glycol, and water (<1mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids.
Specifications

Chemical Identifiers

28395-03-1
364.416
MAEIEVLCKWDQJH-UHFFFAOYSA-N
2471
3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid
C17H20N2O5S
MFCD00078949
bumetanide, 3-butylamino-4-phenoxy-5-sulfamoylbenzoic acid, bumex, burinex, fordiuran, lunetoron, fontego, segurex, bumetanida, bumetanidum
CHEBI:3213
CCCCNC1=C(C(=CC(=C1)C(=O)O)S(=O)(=O)N)OC2=CC=CC=C2

Specifications

White
1g
C17H20N2O5S
14,1484
MAEIEVLCKWDQJH-UHFFFAOYSA-N
3-(butylamino)-4-phenoxy-5-sulfamoylbenzoic acid
2471
364.4
≥98%
230°C to 231°C
28395-03-1
MFCD00078949
bumetanide, 3-butylamino-4-phenoxy-5-sulfamoylbenzoic acid, bumex, burinex, fordiuran, lunetoron, fontego, segurex, bumetanida, bumetanidum
CCCCNC1=C(C(=CC(=C1)C(=O)O)S(=O)(=O)N)OC2=CC=CC=C2
364.416
CHEBI:3213
Solid
Bumetanide
Safety and Handling

Safety and Handling

RTECSNumber : DG4910000