Please login to your online account to display your discounted pricing
Bullard™ Tychem™ SL Hoods for Respirators — Vinyl Lens, 19 in. (48.3cm) Inner Bib
For use with CC20 Airline Respirators and PA20, PA30, PA30IS, EVA, EVAHL PAPRs
$665.52 - $665.52
Specifications
Material Tychem™ SL Coated with Saranex™ 23-P
Material (Lens) Vinyl
Suspension Type No suspension
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
19-065-767
ed bullard company
20SICVN
Double Bib Hood Pack of 5 Each for $665.52
Description & Specifications
Specifications
Material Tychem™ SL Coated with Saranex™ 23-P
Material (Lens) Vinyl
Suspension Type No suspension
Tychem SL is Tychem coated with Saranex™ 23-P to provide protection from an even wider range of chemicals. Hoods cover head, neck, and face; they feature a wide-angled plastic lens.
Double Bib (Inner Bib) Style: Outer bib with a long inner bib; tuck inner bib into shirt or coveralls to funnel excess air from the hood to the body
Double Bib (Inner Bib) Style for Use with Hardhat: Accommodates Bullard hardhats