Bullard™ RT Series Supplied-Air Respirator Breathing Tubes
Breathing tube component for use with RT Series respirators.
$55.95 - $89.27
Specifications
Product Type | Breathing Tube |
---|---|
For Use With (Equipment) | RT Series supplied-air respirators |
Includes | S18051 hose clamp |
Catalog Number | Mfr. No. | Description | Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Color | Price | Quantity | ||||
36-100-3802
|
Bullard™
20BT |
Wire-reinforced, heavy-duty breathing tube | White |
Each for $89.27
|
|
||||
36-100-3803
|
Bullard™
RTBT |
Lightweight, disposable breathing tube | Gray |
Each for $55.95
|
|
||||
Description
- Component for use with RT series respirators
- Choose between heavy-duty or lightweight options
- Breathing tubes must be used in conjunction with hood, flow control device, and air hose
Specifications
Breathing Tube | |
RT Series supplied-air respirators | |
S18051 hose clamp |