Please login to your online account to display your discounted pricing
Bullard™ Hardhat and Cap Faceshield Aluminum Brackets
Rugged brackets are ideal for high-heat industrial applications
$17.62 - $18.84
Specifications
Product Type Gold Line Bracket
Material Aluminum
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Mounting For Use With (Equipment) Product Type Price Quantity    
17-360
ed bullard company
500M
Chemical handling and high heat industrial applications Slot C30 Series cap style hard hat models, faceshield Gold Line Bracket Each for $17.87
Case of 25 Each for $409.00
19-021320
ed bullard company
100G*
Chemical handling and high heat industrial applications Band C30, S61, S62, S51, 911C cap style, 911H Full brim series and bump cap type II hard hat, faceshield Gold Line Bracket Each for $17.62
Case of 25 Each for $403.25
19-021327
ed bullard company
112G
High heat industrial application Band C33, C34, S71 full brim hard hat Gold Line Bracket Each for $18.84
Case of 25 Each for $431.25
Description & Specifications
Specifications
Product Type Gold Line Bracket
Material Aluminum
Aluminum Brackets: Gold Line™ brackets are constructed of rugged, gold anodized aluminum. Highly resistant to corrosion and heat. Ideal for chemical handling and high heat industrial applications. Band- or slot-mounted.