missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Wanzl Bulk Shelving Bottom Cross Bar
Bottom Cross Bar for use with bulk shelving.
$62.85 - $77.89
Specifications
For Use With (Application) | Shelving |
---|---|
Includes | 1 Bottom Cross Bar |
Color | Chrome |
Material | Steel |
Description
- Bottom Cross Bar
- Replacement part for bulk shelving system
Specifications
Shelving | |
1 Bottom Cross Bar | |
Chrome | |
Steel |