Bulbtronics™ Sonntek Deuterium Lamp
Pre-aligned lamp for various models of Agilent™ instruments
$2,398.18 - $2,398.18
Catalog Number | Mfr. No. | For Use With (Equipment) | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Includes | Price | Quantity | ||||
14395358
|
Bulbtronics™
247 |
Model 8453A, 8453E | LH15 holder |
N/A
|
|
||||
Description
- UV/Deuterium
- Comes with holder
Specifications
Deuterium Lamp | |
Deuterium |