Bulbtronics™ General Electric™ Germicidal Bulb
254nm in a variety of styles and dimensions
$36.40 - $186.00
Specifications
Circuitry Preheat
Product Type Linear Double Ended Bulb
Light Source Germicidal
Wavelength 254nm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Base Style Lighting Life Shape Wattage Price Quantity  
Catalog Number Mfr. No. Base Style Lighting Life Shape Wattage Price Quantity  
14395848 Bulbtronics
32550
G5/Mini Bi Pin 6000 hr. T5 Shape 4w Each for $36.40
14395849 Bulbtronics
32558
FA8/slimline, Single Pin 9000 hr. T5 Shape 39w Each for $186.00
14395850 Bulbtronics
32559
G13/Medium Bi Pin 8000 hr. T8 Shape 15w Each for $67.23
14395860 Bulbtronics
32560
G13/Medium Bi Pin 8000 hr. T8 Shape 25w Each for $73.11
Specifications
Description
  • Style: linear double ended
  • Wavelength: 254nm
Description & Specifications
Specifications
Preheat
Linear Double Ended Bulb
Germicidal
254nm