Please login to your online account to display your discounted pricing

Buffer Starter Kit , (500mL Buffers pH 4.0, 7.0, and 10.0 and 125mL Potassium Chloride, 4M), Certified, LabChem™

$133.31 - $133.31

Chemical Identifiers

Chemical Name or Material Buffer Starter Kit
Name Note (500mL Buffers pH 4.0, 7.0, and 10.0 and 125mL Potassium Chloride, 4M)
Purity Grade Certified Grade
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC12100

 
lab chem
LC12100
1 kit Each for $133.31
Description & Specifications

Specifications

Purity Grade Certified Grade
Packaging Poly Bottle
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Quantity 1 kit
Identification Passes Test
Physical Form Liquid
Density 1g/mL
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
Recommended Storage Room Temperature
Name Note (500mL Buffers pH 4.0, 7.0, and 10.0 and 125mL Potassium Chloride, 4M)
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Solubility Soluble in water
Chemical Name or Material Buffer Starter Kit
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May damage fertility or the unborn child.

Room Temperature