missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution, TISAB, Fisher Chemical

For Analysis of the Determination of Fluoride Ion Concentration by Fluoride Ion Electrode

Manufacturer:  Fisher Scientific Company FLSB175-4

 View more versions of this product

Catalog No. SB175-4



 

Specifications Description & Specifications

Specifications

Buffer Solution, TISAB
7647-14-5,127-09-3,482-54-2,9003-39-8,52-51-7
Liquid
0.1ppm max.
Pass Test
Contains 1,2-Cyclohexylene Dinitrilotetraacetic Acid to prevent Aluminum interference.
Clear
Total Ionic Strength Adjustment Buffer
Pass Test
MFCD00146206
Pass Test
Poly Bottle
5.5
4L

Chemical Identifiers

7647-14-5,127-09-3,482-54-2,9003-39-8,52-51-7
MFCD00146206
Buffer Solution, TISAB, Total Ionic Strength Adjustment Buffer

Safety and Handling

SDS & Safety Documents

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0