Buffer Solution, TISAB, Fisher Chemical™
For Analysis of the Determination of Fluoride Ion Concentration by Fluoride Ion Electrode
Supplier: Fisher Chemical SB17520
Specifications
Buffer Solution, TISAB | |
7647-14-5,127-09-3,482-54-2,9003-39-8,52-51-7 | |
5.5 | |
20 L | |
MFCD00146206 |
Total Ionic Strength Adjustment Buffer | |
Undesignated | |
Liquid | |
Pass Test | |
PolyPac™ |