missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer Solution (Phosphate), for Chlorine Determination (DPD Methods), Ricca Chemical

$61.55 - $91.24

Chemical Identifiers

CAS 7778-77-0 (Potassium Phosphate, Monobasic),7558-79-4 (Sodium Phosphate, Dibasic),7487-94-7 (Mercuric Chloride),7732-18-5 (Water, Deionized)
Chemical Name or Material Buffer Solution (Phosphate)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
5805-16 Ricca Chemical Company
RI580516
Poly natural 500mL Each for $61.55
5805-32 Ricca Chemical Company
RI580532
Poly natural 1L Each for $91.24
5805-1 SDS Ricca Chemical Company
RI58051
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7778-77-0 (Potassium Phosphate, Monobasic),7558-79-4 (Sodium Phosphate, Dibasic),7487-94-7 (Mercuric Chloride),7732-18-5 (Water, Deionized)
Buffer Solution (Phosphate)

Specifications

500mL
7732-18-5 (Water, Deionized),7558-79-4 (Sodium Phosphate, Dibasic),7487-94-7 (Mercuric Chloride),7778-77-0 (Potassium Phosphate, Monobasic)
Buffer Solution (Phosphate)
Chlorine Determination (DPD Methods)
Poly natural